Anti-CXCL10 Rabbit Polyclonal Antibody
Catalog # USBII8449-07-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-X-C motif) ligand 10
- Antigen symbol:CXCL10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Neutralisation:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CXCL10 (chemokine (C-X-C motif) ligand 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human