Anti-Interleukin 27 Receptor alpha Rabbit Polyclonal Antibody
Catalog # USBII8443-80B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 27 Receptor Alpha
- Antigen symbol:IL27RA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:611 to 625
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: Interleukin 27 Receptor alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human