Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-Interleukin 27 Receptor alpha Rabbit Polyclonal Antibody
  USBII8443-80
Anti-Interleukin 27 Receptor alpha Rabbit Polyclonal Antibody
  USBII8443-80

 

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 27 Receptor Alpha
  • Antigen symbol:
    IL27RA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,02% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 5-10 ?g/ml

Type: Primary
Antigen: Interleukin 27 Receptor alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human