Anti-Interleukin 18 Receptor alpha Mouse Monoclonal Antibody [clone: 3H2455]
Catalog # USBII8439-95F
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 18 Receptor Alpha
- Antigen symbol:IL18R alpha
- Clonality:Monoclonal
- Clone:3H2455
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Antigen synonyms:CD218a
- Storage buffer:PBS.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Interleukin 18 Receptor alpha
Clonality: monoclonal
Clone: 3H2455
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human