Anti-Interleukin 13 Receptor Mouse Monoclonal Antibody [clone: 9E97]
Catalog # USBII8435-24B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 13 Receptor
- Antigen symbol:IL13R
- Clonality:Monoclonal
- Clone:9E97
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Interleukin 13 Receptor
Clonality: monoclonal
Clone: 9E+97
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human