Anti-Interleukin 12, p40 Rat Monoclonal Antibody [clone: 5C70]
Catalog # USBII8434-30B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 12, p40
- Clonality:Monoclonal
- Clone:5C70
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by ion exchange chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Recommended dilution: ELISA: 1:5000
Type: Primary
Antigen: Interleukin 12, p40
Clonality: monoclonal
Clone: 5C70
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse