Anti-Interleukin 12 Mouse Monoclonal Antibody [clone: 10B2367]
Catalog # USBII8434-06C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 12
- Antigen symbol:IL12
- Clonality:Monoclonal
- Clone:10B2367
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Sheep,Human,Bovine
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:10; ; Western Blot: 1:50-1:100; Functional Assays: 1:100
Type: Primary
Antigen: Interleukin 12
Clonality: monoclonal
Clone: 10B2367
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Human, Sheep