Anti-Interleukin 10 Receptor alpha Rabbit Polyclonal Antibody (PE (Phycoerythrin))
USBII8432-28C-PE
- Antibody type:Primary
- Antigen name:Interleukin 10 Receptor alpha
- Antigen symbol:IL10RA
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by immunoaffinity chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: Interleukin 10 Receptor alpha
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human