Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-Interleukin 10 Receptor alpha Rat Monoclonal Antibody [clone: 10B2192]
  USBII8432-28A
Anti-Interleukin 10 Receptor alpha Rat Monoclonal Antibody [clone: 10B2192]
  USBII8432-28A

 

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 10 Receptor alpha
  • Antigen symbol:
    IL10RA
  • Clonality:
    Monoclonal
  • Clone:
    10B2192
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Mouse
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    250 µg
  • Pk:
    250 µG

 

 

Recommended dilution: Flow Cytometry: 1:10-1:100;

Type: Primary
Antigen: Interleukin 10 Receptor alpha
Clonality: monoclonal
Clone: 10B2192
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Mouse