Anti-Integrin alpha 11 Sheep Polyclonal Antibody
USBII7661-37U1
- Antibody type:Primary
- Antigen name:Integrin Alpha 11
- Antigen symbol:ITGA11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Isotype:IgG
- Reactivity:Human,Mouse
- Form:lyophilized powder
- Amino acid number:19 to 1141
- Storage buffer:PBS,trehalose.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 5-15 ?g/ml
Type: Primary
Antigen: Integrin alpha 11
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse