Anti-Hydroxysteroid Dehydrogenase, 11-beta, Type II Rabbit Polyclonal Antibody
USBIH9117-50R
- Antibody type:Primary
- Antigen name:Hydroxysteroid Dehydrogenase, 11-beta, Type II
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:25 to 40
- Storage buffer:TBS.
- Sequence:RSDLRLGRPLLAALAL
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:150 µl
- Pk:150 µl
Recommended dilution: Western Blot: 1:200
Type: Primary
Antigen: Hydroxysteroid Dehydrogenase, 11-beta, Type II
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat