Anti-17a-Hydroxyprogesterone Mouse Monoclonal Antibody [clone: 9F22]
Catalog # USBIH9111-32A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:17-alpha-Hydroxyprogesterone
- Clonality:Monoclonal
- Clone:9F22
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG3
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: 17a-Hydroxyprogesterone
Clonality: monoclonal
Clone: 9F22
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: