Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody [clone: HL39]
Catalog # USBIH6099-96R
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:HLA Class 2 Antigen DR
- Clonality:Monoclonal
- Clone:HL39
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG3
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, 15mM sodium azide, pH 7,4.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: HLA Class 2 Antigen DR
Clonality: monoclonal
Clone: HL39
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human