Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody [clone: LN-3]
Catalog # USBIH6099-96P
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:HLA Class 2 Antigen DR
- Clonality:Monoclonal
- Clone:LN-3
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Format:Supernatant
- Form:liquid
- Storage buffer:0,7%BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Primary
Antigen: HLA Class 2 Antigen DR
Clonality: monoclonal
Clone: LN-3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human