Order Entry
Czech Republic
ContactUsLinkComponent
Anti-HLA Class 2 Antigen DP, DR Mouse Monoclonal Antibody [clone: HL-40]
Anti-HLA Class 2 Antigen DP, DR Mouse Monoclonal Antibody [clone: HL-40]
Catalog # USBIH6099-85M
Supplier:  US Biological
Anti-HLA Class 2 Antigen DP, DR Mouse Monoclonal Antibody [clone: HL-40]
Catalog # USBIH6099-85M
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HLA Class 2 Antigen DP, DR
  • Clonality:
    Monoclonal
  • Clone:
    HL-40
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Pig,
    Human
  • Form:
    liquid
  • Storage buffer:
    PBS, 15mM sodium azide, pH 7,4.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: HLA Class 2 Antigen DP, DR
Clonality: monoclonal
Clone: HL-40
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Pig