Anti-SLC2A4 Rabbit Polyclonal Antibody
Catalog # USBIG3900-50P
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 2 (facilitated glucose transporter), member 4
- Antigen symbol:Solute carrier family 2, member 4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Recommended dilution: Immunochemistry: 10-20 ?g/ml
Type: Primary
Antigen: SLC2A4 (solute carrier family 2 (facilitated glucose transporter), member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human