Anti-Glucose Regulated Protein 78 Rabbit Polyclonal Antibody
Catalog # USBIG3057-20-200
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Glucose Regulated Protein 78
- Antigen symbol:GRP78
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Hamster,Rabbit,Monkey,Bovine,Xenopus laevis,Rat,Mouse
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: Glucose Regulated Protein 78
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Hamster, Monkey, Mouse, Rabbit, Rat, Xenopus laevis