Anti-Glucose Regulated Protein 75 Mouse Monoclonal Antibody [clone: 10F705]
USBIG3057-12B-25
- Antibody type:Primary
- Antigen name:Glucose Regulated Protein 75
- Antigen symbol:GRP75
- Clonality:Monoclonal
- Clone:10F705
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:25 µg
- Pk:25 µG
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: Glucose Regulated Protein 75
Clonality: monoclonal
Clone: 10F705
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat