Anti-Fibroblast Growth Factor 21 [Cys0] Rabbit polyclonal antibody
Catalog # USBIF4212-93N
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Fibroblast growth factor 21
- Antigen symbol:FGF21
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:lyophilized powder
- Amino acid number:184 to 209
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~1 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Recommended dilution: ELISA: 1:3200
Type: Primary
Antigen: Fibroblast Growth Factor 21 [Cys0]
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human