Anti-FCER1A Mouse monoclonal antibody [clone: AER-37 (CRA1)]
Catalog # USBIF0025-06K
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Fc fragment of IgE, high affinity I, receptor for
- Antigen symbol:FCER1A
- Clonality:Monoclonal
- Clone:AER-37 (CRA1)
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2, 150mM sodium chloride, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 20 ?g/ml
Type: Primary
Antigen: FCER1A (Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide)
Clonality: monoclonal
Clone: AER-37 (CRA1)
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human