Anti-FceRI, alpha Mouse monoclonal antibody [clone: AER-37 (CRA-1)]
Catalog # USBIF0025-06J
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:FCERI, alpha
- Antigen symbol:FCERIA
- Clonality:Monoclonal
- Clone:AER-37 (CRA-1)
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: FceRI, alpha
Clonality: monoclonal
Clone: AER-37 (CRA-1)
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human