Anti-Fas, Neutralizing Mouse Monoclonal Antibody [clone: 0.T.46]
Katalogové číslo USBIF0019-64
Dodavatel: US Biological
Specifikace
- Antibody type:Primary
- Antigen name:Fas, Neutralizing
- Clonality:Monoclonal
- Clone:0.T.46
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Neutralisation:Yes
- Form:liquid
- Storage buffer:PBS, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation and Protein A chormatography
- Size:100 µg
- Pk:100 µG
Specifikace
O tomto produktu
Recommended dilution: Neutralization: 10-500ng/ml; Western Blot: 5 ?g/ml; Flow Cytometry: 5-20 ?g/ml
Type: Primary
Antigen: Fas, Neutralizing
Clonality: monoclonal
Clone: 0.T.46
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human