Order Entry
Czech Republic
ContactUsLinkComponent
Anti-FAS Mouse Monoclonal Antibody [clone: 1.V.2A]
Anti-FAS Mouse Monoclonal Antibody [clone: 1.V.2A]
Catalog # USBIF0019-60B
Supplier:  US Biological
Anti-FAS Mouse Monoclonal Antibody [clone: 1.V.2A]
Catalog # USBIF0019-60B
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Fas cell surface death receptor
  • Antigen symbol:
    FAS
  • Clonality:
    Monoclonal
  • Clone:
    1.V.2A
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    500 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: FAS (Fas cell surface death receptor)
Clonality: monoclonal
Clone: 1.V.2A
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human