Anti-TNFRSF10B Rabbit Polyclonal Antibody
Catalog # USBID9300-52A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:tumor necrosis factor receptor superfamily, member 10b
- Antigen symbol:TNFRSF10B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:250-1:500; Immunochemistry: 5-10 ?g/ml
Type: Primary
Antigen: TNFRSF10B (tumor necrosis factor receptor superfamily, member 10b)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse