Anti-Desmoplakin 1,2 Mouse monoclonal antibody [clone: 10B2716]
USBID3221-91C
- Antibody type:Primary
- Antigen name:Desmoplakin 1,2
- Clonality:Monoclonal
- Clone:10B2716
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Type: Primary
Antigen: Desmoplakin 1,2
Clonality: monoclonal
Clone: 10B2716
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human