Anti-KRT10 Mouse Monoclonal Antibody [clone: 6A400]
Catalog # USBIC9097-09D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:keratin 10
- Antigen symbol:KRT10
- Clonality:Monoclonal
- Clone:6A400
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Pig,Human
- Format:Tissue culture supernatant
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: Immunochemistry: 1:50
Type: Primary
Antigen: KRT10 (keratin 10)
Clonality: Monoclonal
Clone: 6A400
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig