Anti-Cyclic AMP Rabbit Polyclonal Antibody
USBIC8450-05D
- Antibody type:Primary
- Antigen name:Cyclic Adenosine Monophosphate
- Antigen symbol:cAMP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:
- Environmentally Preferable:
- Form:liquid
- Amino acid number:2' to O to
- Storage buffer:PBS, 1% BSA, 0,02% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: EIA: 1:50
Type: Primary
Antigen: Cyclic AMP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: