Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Connexin 30, GJB6, Gap Junction Protein, beta 6, Goat Polyclonal Antibody
Anti-Connexin 30, GJB6, Gap Junction Protein, beta 6, Goat Polyclonal Antibody
  USBIC7853-45
Anti-Connexin 30, GJB6, Gap Junction Protein, beta 6, Goat Polyclonal Antibody
  USBIC7853-45

 

  • Antibody type:
    Primary
  • Antigen name:
    Connexin 30, GJB6, Gap Junction Protein, beta 6,
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    Tris saline, pH 7,2, 0,5% BSA, 0,02% sodium azide.
  • Sequence:
    C-RSKRAQTQKNHPNH
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by peptide affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: ELISA: 1:16000

Type: Primary
Antigen: Connexin 30, GJB6, Gap Junction Protein, beta 6,
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human