Anti-C4BPA Mouse monoclonal antibody [clone: 10-07]
Catalog # USBIC7850-19A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:complement component 4 binding Protein, alpha
- Antigen symbol:C4BPA
- Clonality:Monoclonal
- Clone:10-07
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Form:liquid
- Storage buffer:borate buffered saline, pH 8.4, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: C4BPA (complement component 4 binding protein, alpha)
Clonality: Monoclonal
Clone: 42284
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human