Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Chromosome 20 Open Reading Frame 75 Rabbit polyclonal antibody
Anti-Chromosome 20 Open Reading Frame 75 Rabbit polyclonal antibody
Catalog # USBIC5071-27B
Supplier:  US Biological
Anti-Chromosome 20 Open Reading Frame 75 Rabbit polyclonal antibody
Catalog # USBIC5071-27B
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Chromosome 20 open reading frame 75
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: Chromosome 20 Open Reading Frame 75
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human