Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Chorionic Gonadotropin, Human, beta Mouse Monoclonal Antibody [clone: 9.F.55]
Anti-Chorionic Gonadotropin, Human, beta Mouse Monoclonal Antibody [clone: 9.F.55]
Catalog # USBIC5069-66A
Supplier:  US Biological
Anti-Chorionic Gonadotropin, Human, beta Mouse Monoclonal Antibody [clone: 9.F.55]
Catalog # USBIC5069-66A
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glycoprotein hormones, beta polypeptide
  • Antigen symbol:
    CGB
  • Clonality:
    Monoclonal
  • Clone:
    9.F.55
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    0,9% sodium chloride, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by chromatography
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Type: Primary
Antigen: Chorionic Gonadotropin, Human, beta
Clonality: Monoclonal
Clone: 9.F.55
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human