Anti-Cholinergic Receptor, Nicotinic, alpha 7, Exons 5-10 Goat Polyclonal Antibody
Catalog # USBIC5062-74
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Cholinergic Receptor, Nicotinic, alpha 7, Exons 5-10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Form:liquid
- Storage buffer:Tris, pH 7,3, 0,5% BSA, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA:1:1000
Type: Primary
Antigen: Cholinergic Receptor, Nicotinic, alpha 7, Exons 5-10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human