Anti-Chaperonin 10 Rabbit Polyclonal Antibody
Catalog # USBIC3470-200
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Chaperonin 10
- Antigen symbol:CPN10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Pig,Rabbit,Sheep,Human,Bovine,Guinea pig,Xenopus laevis,Dog,Rat,Mouse
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: Chaperonin 10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Pig, Rabbit, Rat, Sheep, Xenopus laevis