Anti-CES1 Goat polyclonal antibody
Catalog # USBIC3010-02D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Carboxylesterase 1
- Antigen symbol:CES1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Form:liquid
- Storage buffer:TBS, pH 7,3, 0,5% BSA, 0,02% sodium azide.
- Sequence:C-NTQAAQKLKDKE
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: ELISA: 1:8000; Western Blot: 0. 03-0.1 ?g/ml; Immunochemistry: 2.5 ?g/ml
Type: Primary
Antigen: CES1 (carboxylesterase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Human