Anti-CES1 Mouse monoclonal antibody [clone: 11n45]
Katalogové číslo USBIC3010-02C1
Dodavatel: US Biological
Specifikace
- Antibody type:Primary
- Antigen name:Carboxylesterase 1
- Antigen symbol:CES1
- Clonality:Monoclonal
- Clone:11n45
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Form:lyophilized powder
- Amino acid number:19 to 563
- Storage buffer:PBS,trehalose.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µg
- Pk:100 µG
Specifikace
O tomto produktu
Recommended dilution: Western Blot: 2?g/ml; Immunoprecipitation: 25 ?g/ml
Type: Primary
Antigen: CES1 (carboxylesterase 1)
Clonality: Monoclonal
Clone: 11n45
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human