Anti-CTLA4 Hamster monoclonal antibody [clone: UC10-4B9]
Catalog # USBIC2548-83A3
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:cytotoxic T-lymphocyte-associated Protein 4
- Antigen symbol:CTLA4
- Clonality:Monoclonal
- Clone:UC10-4B9
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Hamster
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: CTLA4 (cytotoxic T-lymphocyte-associated protein 4)
Clonality: Monoclonal
Clone: UC10-4B9
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse