Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-KIT Mouse Monoclonal Antibody [clone: 5F6]
  USBIC2448-06D
undefined
Anti-KIT Mouse Monoclonal Antibody [clone: 5F6]
  USBIC2448-06D

 

  • Antibody type:
    Primary
  • Antigen name:
    v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog
  • Antigen symbol:
    KIT
  • Clonality:
    Monoclonal
  • Clone:
    5F6
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    41 to 140
  • Storage buffer:
    PBS, pH 7,2.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: KIT (v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog)
Clonality: Monoclonal
Clone: 5F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human