Anti-KIT Mouse Monoclonal Antibody [clone: 7H14]
Catalog # USBIC2448-04N
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog
- Antigen symbol:KIT
- Clonality:Monoclonal
- Clone:7H14
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Format:ascites
- Form:liquid
- Storage buffer:10mM HEPES, pH 7,5, 150mM sodium chloride,100ug/ml BSA, 50% glycerol and less than 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50
Type: Primary
Antigen: KIT (v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog)
Clonality: Monoclonal
Clone: 7H14
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human