Anti-KIT Mouse Monoclonal Antibody [clone: 5K95]
USBIC2448-04L
- Antibody type:Primary
- Antigen name:v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog
- Antigen symbol:KIT
- Clonality:Monoclonal
- Clone:5K95
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Chicken
- Form:liquid
- Storage buffer:100mM borate buffered saline, pH 8.0. No oramine- containing buffer salts .
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:500 µg
- Pk:500 µG
Type: Primary
Antigen: KIT (v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog)
Clonality: Monoclonal
Clone: 5K95
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Chicken