Anti-KIT Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 7H131]
USBIC2448-03L
- Antibody type:Primary
- Antigen name:v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog
- Antigen symbol:KIT
- Clonality:Monoclonal
- Clone:7H131
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:50mMsodium Phosphate pH 7,5, 500mM Potassium chloride, 150mM NaCl, 15% glycerol, 0,2% BSA, 0,04% NaN3.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:120 Tests
- Pk:120 Tests
Do not freeze!
Type: Primary
Antigen: KIT (v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog)
Clonality: Monoclonal
Clone: 7H131
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human