Order Entry
Czech Republic
ContactUsLinkComponent
Anti-KIT Rat Monoclonal Antibody (Biotin) [clone: 2B8]
Anti-KIT Rat Monoclonal Antibody (Biotin) [clone: 2B8]
  USBIC2448-03C
Anti-KIT Rat Monoclonal Antibody (Biotin) [clone: 2B8]
  USBIC2448-03C

 

  • Antibody type:
    Primary
  • Antigen name:
    v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog
  • Antigen symbol:
    KIT
  • Clonality:
    Monoclonal
  • Clone:
    2B8
  • Conjugation:
    Biotin
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Mouse
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,1% sodium azide
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    500 µg
  • Pk:
    500 µG

 

 

Type: Primary
Antigen: KIT (v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog)
Clonality: Monoclonal
Clone: 2B8
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse