Anti-KIT Rat Monoclonal Antibody [clone: 2B8]
USBIC2448-03AX
- Antibody type:Primary
- Antigen name:v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog
- Antigen symbol:KIT
- Clonality:Monoclonal
- Clone:2B8
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:500 µg
- Pk:500 µG
Type: Primary
Antigen: KIT (v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog)
Clonality: Monoclonal
Clone: 2B8
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse