Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-KIT Rabbit Monoclonal Antibody [clone: 9H219]
  USBIC2448-02P
Anti-KIT Rabbit Monoclonal Antibody [clone: 9H219]
  USBIC2448-02P

 

  • Antibody type:
    Primary
  • Antigen name:
    v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog
  • Antigen symbol:
    KIT
  • Clonality:
    Monoclonal
  • Clone:
    9H219
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Format:
    Supernatant
  • Form:
    liquid
  • Storage buffer:
    50mM Tris-glycine, pH 7,4, 0,15M sodium chloride, 0,01% sodium azide, 0,05% BSA, 40% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:100-250

Type: Primary
Antigen: KIT (v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog)
Clonality: Monoclonal
Clone: 9H219
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human