Anti-Thy1.2 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: 30-H12]
Catalog # USBIC2441-85J
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Thy1.2
- Clonality:Monoclonal
- Clone:30-H12
- Conjugation:PE (Phycoerythrin)/Cy5®
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide and a stabilizing agent.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Thy1.2 (Thy1.2)
Clonality: Monoclonal
Clone: 30-H12
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse