Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CD63 Mouse Monoclonal Antibody [clone: AHN16.1/46-4-5]
Anti-CD63 Mouse Monoclonal Antibody [clone: AHN16.1/46-4-5]
Katalogové číslo USBIC2418-02N
Dodavatel:  US Biological
Anti-CD63 Mouse Monoclonal Antibody [clone: AHN16.1/46-4-5]
Katalogové číslo USBIC2418-02N
Dodavatel:  US Biological

Specifikace

  • Antibody type:
    Primary
  • Antigen name:
    CD63 molecule
  • Antigen symbol:
    CD63
  • Clonality:
    Monoclonal
  • Clone:
    AHN16.1/46-4-5
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    50mMsodium phosphate, pH 7,5, 100mM potassium chloride, 150mM sodium chloride.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifikace

O tomto produktu

Type: Primary
Antigen: CD63 (CD63 molecule)
Clonality: Monoclonal
Clone: AHN16.1/46-4-5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human