Anti-CD59 Glycoprotein Mouse Monoclonal Antibody [clone: MEM-43]
USBIC2413-22G
- Antibody type:Primary
- Antigen symbol:CD59 Glycoprotein
- Clonality:Monoclonal
- Clone:MEM-43
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:50 µg
- Pk:50 µG
Recommended dilution: Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: CD59 Glycoprotein
Clonality: Monoclonal
Clone: MEM-43
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human