Anti-CD55 Mouse Monoclonal Antibody [clone: MEM-118]
Catalog # USBIC2410-10G
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:CD55 molecule, decay accelerating factor for complement (Cromer blood group)
- Antigen symbol:CD55
- Clonality:Monoclonal
- Clone:MEM-118
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgM
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, 15mM sodium azide, pH 7,4.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by size exclusion chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: CD55 (CD55 molecule, decay accelerating factor for complement (Cromer blood group))
Clonality: Monoclonal
Clone: MEM-118
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human