Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-CD34 Mouse Monoclonal Antibody [clone: QBEND-10]
Catalog # USBIC2386-31F
Supplier:  US Biological
undefined
Anti-CD34 Mouse Monoclonal Antibody [clone: QBEND-10]
Catalog # USBIC2386-31F
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD34 molecule
  • Antigen symbol:
    CD34
  • Clonality:
    Monoclonal
  • Clone:
    QBEND-10
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Immunochemistry: 1:800

Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: Monoclonal
Clone: QBEND-10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human