Anti-ITGB2 Mouse Monoclonal Antibody [clone: MEM-48]
Katalogové číslo USBIC2269-08J
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:integrin, beta 2 (complement component 3 receptor 3 and 4 subunit)
- Antigen symbol:ITGB2
- Clonality:Monoclonal
- Clone:MEM-48
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 15mM sodium azide, pH 7,4.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 2?g/ml; Western Blot: 2?g/ml Immunochemistry: 10 ?g/ml; Immunoprecipitation: 2?g/ml
Type: Primary
Antigen: ITGB2 (integrin, beta 2 (complement component 3 receptor 3 and 4 subunit))
Clonality: Monoclonal
Clone: MEM-48
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human