Order Entry
Czech Republic
ContactUsLinkComponent
Anti-ITGB2 Mouse Monoclonal Antibody [clone: TS1/18]
Anti-ITGB2 Mouse Monoclonal Antibody [clone: TS1/18]
Catalog # USBIC2269-08F
Supplier:  US Biological
Anti-ITGB2 Mouse Monoclonal Antibody [clone: TS1/18]
Catalog # USBIC2269-08F
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    integrin, beta 2 (complement component 3 receptor 3 and 4 subunit)
  • Antigen symbol:
    ITGB2
  • Clonality:
    Monoclonal
  • Clone:
    TS1/18
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Pig,
    Human
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: ITGB2 (integrin, beta 2 (complement component 3 receptor 3 and 4 subunit))
Clonality: Monoclonal
Clone: TS1/18
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Pig